missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLE3 Antibody (CL3573), Novus Biologicals™
Mouse Monoclonal Antibody
£222.00 - £443.00
Specifications
| Antigen | TLE3 |
|---|---|
| Clone | CL3573 |
| Dilution | Western Blot 1 μg/mL, Immunohistochemistry 1:10000 - 1:20000, Immunocytochemistry/Immunofluorescence 2-10 μg/mL, Immunohistochemistry-Paraffin 1:10000 - 1:20000, KnockDown Validated |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18294973
|
Novus Biologicals
NBP2-61621 |
100 μL |
£443.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18681969
|
Novus Biologicals
NBP2-61621-25ul |
25 μL |
£222.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TLE3 Monoclonal specifically detects TLE3 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
| TLE3 | |
| Western Blot 1 μg/mL, Immunohistochemistry 1:10000 - 1:20000, Immunocytochemistry/Immunofluorescence 2-10 μg/mL, Immunohistochemistry-Paraffin 1:10000 - 1:20000, KnockDown Validated | |
| Unconjugated | |
| Mouse | |
| Human | |
| 7090 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: GSQAHLTVKDEKNHHELDHRERESSANNSVSPSESLRASEKHR | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| CL3573 | |
| Monoclonal | |
| Purified | |
| Signal Transduction | |
| ESG3Enhancer of split groucho-like protein 3, ESGenhancer of split groucho 3, GRG3, HsT18976, KIAA1547FLJ39460, transducin-like enhancer of split 3 (E(sp1) homolog, Drosophila), transducin-like enhancer of split 3, homolog of Drosophila E(sp1), transducin-like enhancer protein 3 | |
| TLE3 | |
| IgG1 | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title