missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TLE2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16647624
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16647624 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16647624 Supplier Invitrogen™ Supplier No. PA5101145

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total TLE2.

TLE2 (Transducin Like Enhancer Of Split 2) is a Protein Coding gene. Among its related pathways are Signaling by Wnt and Wnt / Hedgehog / Notch. Gene Ontology (GO) annotations related to this gene include transcription corepressor activity. An important paralog of this gene is TLE1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TLE2
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene TLE2
Gene Accession No. Q04725
Gene Alias enhancer of split groucho-like protein 2; ESG; ESG2; ESG-2; GRG2; TLE2; transducin like enhancer of split 2; transducin-like enhancer of split 2 (E(sp1) homolog, Drosophila); transducin-like enhancer of split 2, homolog of Drosophila E(sp1); transducin-like enhancer protein 2
Gene Symbols TLE2
Host Species Rabbit
Immunogen A synthesized peptide derived from human TLE2(Accession Q04725), corresponding to amino acid residues P194-P244.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7089
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.