Learn More
TLE1 Mouse, anti-Human, CoraLite Plus 488, Clone: 2F2H11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-66216-20UL
Description
The TLE1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TLE1. This antibody recognizes human antigen. The TLE1 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| TLE1 | |
| 2F2H11 | |
| CoraLite Plus 488 | |
| Mouse | |
| Primary | |
| Human | |
| 155 kDa | |
| IgG2b |
| Immunofluorescence | |
| 1000 μg/mL | |
| E(Sp1) homolog, ESG, ESG1, GRG1, TLE1 | |
| 20 μL | |
| 7088 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |