missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLE1 Antibody (TLE1/2051), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79913AF647
This item is not returnable.
View return policy
Description
TLE1 Monoclonal specifically detects TLE1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| TLE1 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| TLE1 | |
| Recombinant human TLE1 fragment (aa 175-338) (exact sequence is proprietary) (Uniprot: Q04724) | |
| 0.1 mL | |
| Signal Transduction | |
| Human | |
| IgG2a κ |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| TLE1/2051 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| E(Sp1) homolog, enhancer of split groucho 1, ESG, ESG1Enhancer of split groucho-like protein 1, GRG1, transducin-like enhancer of split 1 (E(sp1) homolog, Drosophila), transducin-like enhancer of split 1, homolog of Drosophila E(sp1), transducin-like enhancer protein 1 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction