missing translation for 'onlineSavingsMsg'
Learn More

TLE Pan Polyclonal Antibody

Product Code. 11546061
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11546061 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11546061 Supplier Invitrogen Supplier No. PA517241

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

TLE Pan Polyclonal Antibody

TLE Pan Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen TLE Pan
Applications Western Blot
Classification Polyclonal
Concentration 261 μg/mL
Conjugate Unconjugated
Formulation 0.01M HEPES with 100μg/mL BSA, 50% glycerol, 0.15M NaCl and no preservative; pH 7.5
Gene tle2
Gene Accession No. Q04724, Q04725, Q04726, Q04727, Q08122, Q62440, Q62441, Q9WVB2
Gene Alias 2610103N05Rik; 5730411M05Rik; AA792082; B lymphocyte gene 1; BCE1; Bce-1; C230057C06Rik; E(Sp1) homolog; E(spI); enhancer of split groucho; enhancer of split groucho 3; enhancer of split groucho 4; enhancer of split groucho-like protein 1; Enhancer of split groucho-like protein 2; enhancer of split groucho-like protein 3; Esg; esg1; ESG-1; ESG2; ESG-2; esg3; ESG-3; ESG4; ESG-4; Esp2; Esp3; ESTM13; Estm14; fc18h03; GRG1; Grg-1; GRG2; GRG3; Grg-3; Grg3a; Grg3b; GRG4; Grg-4; gro; groucho protein; groucho-related gene 4; groucho-related protein; groucho-related protein 1; Groucho-related protein 4; HsT18976; KIAA1261; KIAA1547; mKIAA1547; mKIAA4188; Protein ESP2; rTLE3; sb:eu1034; sb:eu408; si:ch211-81i17.1; si:dkey-42p8.3; TLE family member 2, transcriptional corepressor a; TLE family member 2, transcriptional corepressor b; tle1; tle1.S; Tle2; tle2.L; tle2a; tle2b; TLE3; tle3a; tle3b; tle4; tle4.S; Tle4l; transducin like enhancer of split 1; transducin like enhancer of split 1 S homeolog; transducin like enhancer of split 2; transducin like enhancer of split 2 L homeolog; transducin like enhancer of split 2a; transducin like enhancer of split 2b; transducin like enhancer of split 3; transducin like enhancer of split 4; transducin like enhancer of split 4 S homeolog; transducin-like enhancer of split 1 (E(sp1) homolog); transducin-like enhancer of split 1 (E(sp1) homolog, Drosophila); transducin-like enhancer of split 1, homolog of Drosophila E(spl); transducin-like enhancer of split 2; transducin-like enhancer of split 2 (E(sp1) homolog); transducin-like enhancer of split 2 (E(sp1) homolog, Drosophila); transducin-like enhancer of split 2 homolog; transducin-like enhancer of split 2, homolog of Drosophila E(sp1); transducin-like enhancer of split 2, homolog of Drosophila E(spl); transducin-like enhancer of split 2; transducin-like enhancer protein 2a; transducin-like enhancer of split 3; transducin-like enhancer of split 3 (E(sp1) homolog) a; transducin-like enhancer of split 3 (E(sp1) homolog) b; transducin-like enhancer of split 3 (E(sp1) homolog, Drosophila); transducin-like enhancer of split 3 homolog of Drosophila; transducin-like enhancer of split 3 short isoform; transducin-like enhancer of split 3, E(spl) homolog; transducin-like enhancer of split 3, homolog of Drosophila E(sp1); transducin-like enhancer of split 3, homolog of Drosophila E(spl); transducin-like enhancer of split 4; transducin-like enhancer of split 4 (E(sp1) homolog); transducin-like enhancer of split 4 (E(sp1) homolog, Drosophila); transducin-like enhancer of split 4 (homolog of Drosophila E(spl)) - like; transducin-like enhancer of split 4, E(spl) homolog; transducin-like enhancer of split 4, homolog of Drosophila E(sp1); transducin-like enhancer of split 4, homolog of Drosophila E(spl); transducin-like enhancer protein 1; Transducin-like enhancer protein 2; transducin-like enhancer protein 3; Transducin-like enhancer protein 4; wu:fb96a10; wu:fc18h03; X83332; X83333; XELAEV_18001139mg; XELAEV_18006642mg; XELAEV_18010139mg; Xesg1; xgrg1; xgrg3; Xgrg4; XGrg-4; X-groucho; XTCF-4; Xtle4
Gene Symbols Tle1, tle2, TLE3, TLE4
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues surrounding Asp760 of human TLE1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 21885, 21886, 21887, 21888, 7088, 7089, 7090, 7091
Target Species Human, Mouse, Monkey, Drosophila, Rat, X. laevis, Zebrafish
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.