missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TL1A/TNFSF15 Rabbit anti-Human, Clone: VEGI/2052R, Novus Biologicals™
Rabbit Monoclonal Antibody
£235.00
Specifications
| Antigen | TL1A/TNFSF15 |
|---|---|
| Clone | VEGI/2052R |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
| Applications | Immunohistochemistry (Paraffin), Peptide Array |
Description
TL1A/TNFSF15 Monoclonal specifically detects TL1A/TNFSF15 in Human samples. It is validated for Immunohistochemistry-Paraffin, Protein Array.Specifications
| TL1A/TNFSF15 | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin), Peptide Array | |
| Unconjugated | |
| Rabbit | |
| Apoptosis, Biologically Active Proteins, Cell Cycle and Replication | |
| 10 mM PBS with 0.05% BSA | |
| 9966 | |
| IgG | |
| Protein A purified |
| VEGI/2052R | |
| Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| MGC129934, MGC129935, TL1A, TL1Vascular endothelial cell growth inhibitor, TNF superfamily ligand TL1A, tumor necrosis factor (ligand) superfamily, member 15, tumor necrosis factor ligand superfamily member 15, vascular endothelial growth inhibitor-192A, VEGI192A, VEGITNF ligand-related molecule 1 | |
| Recombinant full-length human TL1A/TNFSF15 protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title