missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TL1A/TNFSF15 Antibody (VEGI/1283), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-53388-20UG
This item is not returnable.
View return policy
Description
TL1A/TNFSF15 Monoclonal specifically detects TL1A/TNFSF15 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| TL1A/TNFSF15 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot 1 - 2 ug/ml, Flow Cytometry 0.5 - 1 ug/million cells, Immunocytochemistry/Immunofluorescence 1 - 2 ug/ml, Immunohistochemistry-Paraffin 1 - 2 ug/ml | |
| MGC129934, MGC129935, TL1A, TL1Vascular endothelial cell growth inhibitor, TNF superfamily ligand TL1A, tumor necrosis factor (ligand) superfamily, member 15, tumor necrosis factor ligand superfamily member 15, vascular endothelial growth inhibitor-192A, VEGI192A, VEGITNF ligand-related molecule 1 | |
| Mouse | |
| 22 kDa | |
| 20 μg | |
| Apoptosis, Biologically Active Proteins, Cell Cycle and Replication | |
| 9966 | |
| Store at 4C. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| VEGI/1283 | |
| Unconjugated | |
| 10mM PBS with 0.05% BSA with 0.05% Sodium Azide | |
| TNFSF15 | |
| Recombinant human full-length VEGI fragment | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction