missing translation for 'onlineSavingsMsg'
Learn More

TL1A/TNFSF15 Antibody (VEGI/1283), DyLight 350, Novus Biologicals™

Product Code. 18407984 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18407984 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18407984 Supplier Novus Biologicals Supplier No. NBP254513UV

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TL1A/TNFSF15 Monoclonal antibody specifically detects TL1A/TNFSF15 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF
TRUSTED_SUSTAINABILITY

Specifications

Antigen TL1A/TNFSF15
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), CyTOF
Classification Monoclonal
Clone VEGI/1283
Conjugate DyLight 350
Formulation 50 mM Sodium Borate
Gene Alias MGC129934, MGC129935, TL1A, TL1Vascular endothelial cell growth inhibitor, TNF superfamily ligand TL1A, tumor necrosis factor (ligand) superfamily, member 15, tumor necrosis factor ligand superfamily member 15, vascular endothelial growth inhibitor-192A, VEGI192A, VEGITNF ligand-related molecule 1
Host Species Mouse
Immunogen Recombinant human TL1A/TNFSF15 fragment (Uniprot: O95150)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, BiologicAll speciesy Active Proteins, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 9966
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.