missing translation for 'onlineSavingsMsg'
Learn More

TL1A/TNFSF15 Antibody (TAN2-2), Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18377780 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18377780 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18377780 Supplier Novus Biologicals Supplier No. NBP281120AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

TL1A/TNFSF15 Monoclonal specifically detects TL1A/TNFSF15 in Mouse samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TL1A/TNFSF15
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone TAN2-2
Conjugate Alexa Fluor 350
Dilution Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias MGC129934, MGC129935, TL1A, TL1Vascular endothelial cell growth inhibitor, TNF superfamily ligand TL1A, tumor necrosis factor (ligand) superfamily, member 15, tumor necrosis factor ligand superfamily member 15, vascular endothelial growth inhibitor-192A, VEGI192A, VEGITNF ligand-related molecule 1
Gene Symbols TNFSF15
Host Species Rabbit
Immunogen This antibody was raised by immunising rats with a soluble recombinant TL1A/TNFSF15 (T77-L252):Fc protein.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Biologically Active Proteins, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 9966
Test Specificity This antibody is specific for TL1A/TNFSF15, a member of the TNF ligand family. TL1A/TNFSF15 is expressed by endothelial cells, tissue macrophages, monocyte-derived dendritic cells, plasma cells and lamina propria-derived lymphocytes. Among peripheral blood cells, TL1A/TNFSF15 has been shown to be expressed predominantly by CD4+CCR9+ lymphocytes. Its expression has been shown to be induced by TNF alpha, IL-1 alpha and IFN gamma.
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.