missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ TL 13-12
GREENER_CHOICE

Product Code. 18706246
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
This item is not returnable. View return policy

Product Code. 18706246

Brand: Tocris Bioscience™ 6744/5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective ALK Degrader (PROTAC™)

Selective anaplastic lymphoma kinase (ALK) degrader (DC50 values are 10 and 180 nM in H3122 and Karpas 299 cells, respectively). Comprises the cereblon E3 ligase ligand Pomalidomide (Cat. No. 6302), conjugated to an ALK inhibitor. Inhibits proliferation of ALK-positive cancer cell lines. Exhibits higher selectivity for ALK over Aurora A kinase compared with TL 13 -112 (Cat.No. 6745). Maximum degradation is exhibited at 16 h.

Negative control TL 13-22 (Cat. No. 6747) also available.

TL 13-12 is a selective anaplastic lymphoma kinase (ALK) Degrader (PROTAC®) (DC50 values are 10 and 180 nM in H3122 and Karpas 299 cells, respectively). Comprises the cereblon E3 ligase ligand Pomalidomide (Cat. No. 6302), conjugated to an ALK inhibitor. Inhibits proliferation of ALK-positive cancer cell lines. Exhibits higher selectivity for ALK over Aurora A kinase compared with TL 13 -112 (Cat.No. 6745). Maximum degradation is exhibited at 16 h.

Negative control TL 13-22 (Cat. No. 6747) also available.

PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-(2-(2-(2-(4-(4-((5-Chloro-4-((2-(isopropylsulfonyl)phenyl)amino)pyrimidin-2-yl)amino)-3-methoxyphenyl)piperazin-1-yl)ethoxy)ethoxy)ethyl)-2-((2-(2,6-dioxopiperidin-3-yl)-1,3-dioxoisoindolin-4-yl)amino)acetamide
CAS 2229037-04-9
Quantity 5 mg
Molecular Formula C45H53ClN10O10S
Formula Weight 961.48
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.