missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ TL 0259
GREENER_CHOICE

Product Code. p-7182958
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18766816

Brand: Tocris Bioscience™ 6981/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective and highly potent Fgr and Lyn kinase inhibitor; orally bioavailable

TL 0259 is a selective and highly potent Fgr and Lyn inhibitor (IC50 values are 0.03 and 0.1 nM, respectively). Exhibits selectivity for Fgr and Lyn over other Src family kinases and other nonreceptor and receptor tyrosine kinases. Suppresses proliferation of acute myelogenous leukemia (AML) cell lines in vitro. Eliminates leukemic cells from spleen and bone of mice bearing AML xenografts. Orally bioavailable.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-[(6,7-Dimethoxy-4-quinazolinyl)oxy]-N-[4-[(4-ethyl-1-piperazinyl)methyl]-3-(trifluoromethyl)phenyl]-4-methylbenzamide dihydrochloride
Quantity 10 mg
Molecular Formula C32H34F3N5O4.2HCl
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.