missing translation for 'onlineSavingsMsg'
Learn More

TKTL1 Antibody (1D6), Novus Biologicals™

Product Code. 18319028 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18319028 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18319028 Supplier Novus Biologicals Supplier No. H00008277M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TKTL1 Monoclonal antibody specifically detects TKTL1 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen TKTL1
Applications Western Blot, ELISA, Immunocytochemistry, Sandwich ELISA
Classification Monoclonal
Clone 1D6
Conjugate Unconjugated
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH25382
Gene Alias TK 2, TKR, TKR EC 2.2.1.1, TKT2Transketolase 2, transketolase-2, transketolase-like 1, transketolase-like protein 1, Transketolase-related protein
Host Species Mouse
Immunogen TKTL1 (AAH25382, 1 a.a. ~ 596 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MADAEARAEFPEEARPDRGTLQVFQDMASRLRIHSIRATCSTSSGHPTSCSSSSEIMSVLFFYIMRYKQSDPENPDNDRFVLAKRLSFVDVATGWLGQGLGVACGMAYTGKYFDRASYRVFCLMSDGESSEGSVWEAMAFASYYSLDNLVATFDVNRLGHSGALPAEHCINIYQRRCEAFGWNTYVVDGRDVEALCQVFWQASQVKHKPTAVVAKTFKGRGTPSIEDAESWHAKPMPRERADAIIKLIESQIQTSRNLDPQPPIEDSPEVNITDVRMTSPPDYRVGDKIATRKACGLALAKLGYANNRVVVLDGDTRYSTFSEIFNKEYPERFIECFMAEQNMVSVALGCASRGRTIAFASTFAAFLTRAFDHIRIGGLAESNINIIGSHCGVSVGDDGASQMALEDIAMFRTIPKCTIFYPTDAVSTEHAVALAANAKGMCFIRTTRPETMVIYTPQERFEIGQAKVLRHCVSDKVTVIGAGITVYEALAAADELSKQDIFIRVIDLFTIKPLDVATIVSSAKATEGRIITVEDHYPQGGIGEAVCAAVSMDPDIQVHSLAVSGVPQSGKSEELLDMYGISARHIIVAVKCMLLN
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8277
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.