missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TK2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA513813
This item is not returnable.
View return policy
Description
TK2 Polyclonal Antibody for Western Blot
Thymidine kinase-2 (TK2) is a deoxyribonucleoside kinase that phosphorylates thymidine, deoxycytidine, and deoxyuridine, and also phosphorylates antiviral and anticancer nucleoside analogs. Both recombinant and native forms of the enzyme have broad substrate specificity and complex kinetics, suggesting that it may play a role in the activation of chemotherapeutic nucleoside analogs. The highest levels of expression are observed in testis and ovary. The main supply of deoxyribonucleotides (dNTPs) for mitochondrial DNA synthesis comes from the salvage pathway initiated by deoxyguanosine kinase (DGK) and TK2. The association of mitochondrial DNA depletion with mutations in the genes encoding these 2 kinases suggests that the salvage pathway enzymes are involved in the maintenance of balanced mitochondrial dNTP pools.
Specifications
| TK2 | |
| Polyclonal | |
| Unconjugated | |
| Tk2 | |
| AU024611; MTDPS2; MTTK; Mt-TK; SCA31; thymidine kinase 2, mitochondrial; thymidine kinase 2; thymidine kinase 2, mitochondrial; TK2 | |
| Rabbit | |
| Ammonium Sulfate Precipitation, Size-exclusion - Dialysis | |
| RUO | |
| 57813, 7084 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1.6 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| O00142, Q9R088 | |
| Tk2 | |
| KLH conjugated synthetic peptide between 8-38 amino acids from the N-terminal region of human Thymidine Kinase 2. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction