missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tizoxanide, Mikromol
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
Tizoxanide, Mikromol
Specifications
Specifications
| Chemical Name or Material | Tizoxanide |
| CAS | 173903-47-4 |
| Analyte or Component Names | Tizoxanide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C10 H7 N3 O4 S |
| Certifications/Compliance | ISO 9001 |
| InChI Formula | InChI=1S/C10H7N3O4S/c14-7-4-2-1-3-6(7)9(15)12-10-11-5-8(18-10)13(16)17/h1-5,14H,(H,11,12,15) |
| Solution Type | Neat |
| SMILES | Oc1ccccc1C(=O)Nc2ncc(s2)[N+](=O)[O-] |
| IUPAC Name | 2-hydroxy-N-(5-nitro-1,3-thiazol-2-yl)benzamide |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction