missing translation for 'onlineSavingsMsg'
Learn More

Tizoxanide, MedChemExpress

Product Code. 16495288
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16495288

Brand: Medchem Express HY12687

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Tizoxanide (TIZ) is the active metabolite of Nitazoxanide, which is a thiazolide anti-infective compound against anaerobic bacteria, protozoa, and a range of viruses.

Tizoxanide (TIZ) has anti-HIV-1 activities and potent inhibition of both HBV and HCV replication with values EC50 of 0.46μM and 0.15 μM, respectively. Tizoxanide also exerts anti-inflammatory effects by inhibiting the production of pro-inflammatory cytokines and suppressing of the activation of the NF-κB and the MAPK signaling pathways in LPS-treated macrophage cells.

Specifications

Chemical Name or Material Tizoxanide
Physical Form Solid
CAS 173903-47-4
Color Light Yellow
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C10H7N3O4S
For Use With (Application) COVID-19-anti-virus
Synonym TIZ
Solubility Information DMSO : ≥ 38 mg/mL (143.26 mM)
SMILES O=C(NC1=NC=C([N+]([O-])=O)S1)C2=CC=CC=C2O
Molecular Weight (g/mol) 265.25
Formula Weight 265.25
Percent Purity 98.1%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.