missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tizoxanide-d4, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Tizoxanide-d4 |
| CAS | 1246817-56-0 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C10H3D4N3O4S |
| Quantity | 10 mg |
| Synonym | 2-Hydroxy-N-(5-nitro-2-thiazolyl)benzamide-d4, NSC 697856-d4, TIZ-d4, Desacetylnitazoxanide-d4 |
| InChI Formula | InChI=1S/C12H9N3O5S/c1-7(16)20-9-5-3-2-4-8(9)11(17)14-12-13-6-10(21-12)15(18)19/h2-6H,1H3,(H,13,14,17)/i2D,3D,4D,5D |
| SMILES | OC1=C([2H])C([2H])=C([2H])C([2H])=C1C(NC2=NC=C([N+]([O-])=O)S2)=O |
| IUPAC Name | [2,3,4,5-tetradeuterio-6-[(5-nitro-1,3-thiazol-2-yl)carbamoyl]phenyl] acetate |
| Molecular Weight (g/mol) | 269.27 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction