missing translation for 'onlineSavingsMsg'
Learn More

Tizaterkib, MedChemExpress

Product Code. 16434468
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16434468

Brand: Medchem Express HY111483

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Tizaterkib (AZD0364) is a potent and selective ERK2 inhibitor extracted from patent WO2017080979A1, compound example 18, has an IC50 of 0.6 nM.

Tizaterkib (AZD0364) is a potent and selective ERK2 inhibitor extracted from patent WO2017080979A1, compound example 18, has an IC50 of 0.6 nM.

Specifications

Chemical Name or Material Tizaterkib
CAS 2097416-76-5
Color White
Purity Grade Notes Research
Molecular Formula C24H24F2N8O2
Quantity 10 mg
Synonym AZD0364
Solubility Information DMSO : ≥ 100 mg/mL (202.22 mM)
SMILES O=C1C2=NC(C3=NC(NC4=CC=NN4C)=NC=C3C)=CN2C[C@H](COC)N1CC5=CC=C(F)C(F)=C5
Molecular Weight (g/mol) 494.5
Formula Weight 494.5
Percent Purity 99.31%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.