missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tizaterkib, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
Description
Tizaterkib (AZD0364) is a potent and selective ERK2 inhibitor extracted from patent WO2017080979A1, compound example 18, has an IC50 of 0.6 nM.
Specifications
Specifications
| Chemical Name or Material | Tizaterkib |
| CAS | 2097416-76-5 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C24H24F2N8O2 |
| Quantity | 25 mg |
| Synonym | AZD0364 |
| Solubility Information | DMSO : ≥ 100 mg/mL (202.22 mM) |
| SMILES | O=C1C2=NC(C3=NC(NC4=CC=NN4C)=NC=C3C)=CN2C[C@H](COC)N1CC5=CC=C(F)C(F)=C5 |
| Molecular Weight (g/mol) | 494.5 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction