missing translation for 'onlineSavingsMsg'
Learn More

Tizanidine Hydrochloride, TRC

Product Code. 30453582
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30453582 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30453582 Supplier Toronto Research Chemicals Supplier No. TRCT44950010MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 64461-82-1
Molecular Formula C9 H8 Cl N5 S . Cl H
Quantity 10 mg
Synonym 5-Chloro-N-(4,5-dihydro-1H-imidazol-2-yl)-2,1,3-benzothiadiazol-4-amine monohydrochloride, 5-Chloro-4-(2-imidazolin-2-ylamino)-2,1,3-benzothiadiazole hydrochloride, AB 021, AN 021, DS 103-282, Sirdalud, Ternelin, Tizan, Tizanidine Hydrochloride, Tizpa, Zanaflex
InChI Formula InChI=1S/C9H8ClN5S.ClH/c10-5-1-2-6-8(15-16-14-6)7(5)13-9-11-3-4-12-9;/h1-2H,3-4H2,(H2,11,12,13);1H
SMILES Cl.Clc1ccc2nsnc2c1NC3=NCCN3
IUPAC Name 5-chloro-N-(4,5-dihydro-1H-imidazol-2-yl)-2,1,3-benzothiadiazol-4-amine;hydrochloride
Molecular Weight (g/mol) 290.17
Formula Weight 288.9956
Chemical Name or Material Tizanidine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.