missing translation for 'onlineSavingsMsg'
Learn More

Tizanidine hydrochloride, Thermo Scientific™

Product Code. 17790679 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 17790679

Brand: Thermo Scientific Acros 470000250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

Chemical Name or Material Tizanidine hydrochloride
CAS 64461-82-1
Synonym 5-Chloro-4-[(4,5-dihydro-1 H-imidazol-2-yl)amino]-2,1,3-benzothiadiazole hydrochloride
Quantity 25 g
Solubility Soluble aqueous solution
Melting Point 280°C
Shipping Condition No special requirements
Product Line Life science
Molecular Formula C9 H8 Cl N5 S . H Cl
Physical Form Powder
Color White to Yellow to Light-brown
Purity ≥97.5%
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.