missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tizanidine hydrochloride, Thermo Scientific™
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Specifications
Specifications
| Chemical Name or Material | Tizanidine hydrochloride |
| CAS | 64461-82-1 |
| Synonym | 5-Chloro-4-[(4,5-dihydro-1 H-imidazol-2-yl)amino]-2,1,3-benzothiadiazole hydrochloride |
| Quantity | 5 g |
| Solubility | Soluble aqueous solution |
| Melting Point | 280°C |
| Shipping Condition | No special requirements |
| Product Line | Life science |
| Molecular Formula | C9 H8 Cl N5 S . H Cl |
| Physical Form | Powder |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction