missing translation for 'onlineSavingsMsg'
Learn More

Tizanidine hydrochloride, MedChemExpress

Product Code. 16488166
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16488166 100 mg 100mg
16428408 10 mg 10mg
16404297 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16488166 Supplier Medchem Express Supplier No. HYB0194A
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Tizanidine hydrochloride is an α2-adrenergic receptor agonist and inhibits neurotransmitter release from CNS noradrenergic neurons.

Tizanidine hydrochloride is an α2-adrenergic receptor agonist and inhibits neurotransmitter release from CNS noradrenergic neurons.

Specifications

Chemical Name or Material Tizanidine hydrochloride
Physical Form Solid
CAS 64461-82-1
Color Light Yellow
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C9H9Cl2N5S
For Use With (Application) Neuroscience-Neuromodulation
Solubility Information H2O : 20 mg/mL (68.93 mM; Need ultrasonic)
SMILES ClC1=C(NC2=NCCN2)C3=NSN=C3C=C1.Cl
Molecular Weight (g/mol) 290.17
Formula Weight 290.17
Percent Purity 98.66%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.