missing translation for 'onlineSavingsMsg'
Learn More

Tivantinib, MedChemExpress

Product Code. 16461316
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
500 mg
Unit Size:
100mg
10mg
200mg
500mg
50mg
5mg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16461316 200 mg 200mg
16460548 5 mg 5mg
16418088 10 mg 10mg
16417856 100 mg 100mg
16404007 50 mg 50mg
16481916 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 16461316 Supplier Medchem Express Supplier No. HY50686
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Tivantinib is a highly selective c-Met tyrosine kinase inhibitor with a Ki of 355 nM.

Tivantinib is a highly selective c-Met tyrosine kinase inhibitor with a Ki of 355 nM.

Specifications

Chemical Name or Material Tivantinib
CAS 905854-02-6
Color Brown
Purity Grade Notes Research
Molecular Formula C23H19N3O2
Quantity 200 mg
Synonym ARQ 197
Solubility Information DMSO : ≥ 100 mg/mL (270.69 mM)
SMILES O=C(NC1=O)[C@@H](C2=CN3C4=C(C=CC=C42)CCC3)[C@@H]1C5=CNC6=C5C=CC=C6
Molecular Weight (g/mol) 369.42
Formula Weight 369.42
Percent Purity 96.99%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.