missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- pH/mV-input with 24 bit transducer Electrode socket according to DIN 19 262 or additional with BNC socket insert RFID receiver for SI Analytics ID electrodes
- Connector (μA) for double platinum electrodes Polarisation voltage adjustable from 40 to 220 mV
- Highly visible full color display, that can be easily viewed from a distance and extreme angles With new interchangeable modules which all relevant reagent and unit data can be stored
- Expandable thanks to the 2 x USB-host, 1 x USB-PC, 1 x LAN and 2 x RS232 ports
- Connectable are e.g. USB keyboard, USB printer, barcode reader, USB flash drives, balances, PC and further devices such as piston burettes and sample changers
- Storage of results via USB or LAN connection, including method transfer
Specifications
Specifications
| Includes | Stand, TZ 1510, electrode clamp Z 305, hand controller TZ 3880 |
| Material | Polypropylene |
| Model | TitroLine 7750 |
| Range (pH) | -3 to 18 |
| Temperature Range (Metric) Ambient | 10°C to 40°C |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
