missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ QuantiGene™ Plex Automation Setup Kit

Product Code. 32045022
Change view
Click to view available options
Quantity:
6 Tests
Unit Size:
6 tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
32045022 6 Tests 6 tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32045022 Supplier Invitrogen™ Supplier No. QGPSETUP384
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The QuantiGene Plex Automation Setup Kit enables setup, testing, and validation of instruments to automate crucial steps of the QuantiGene Plex 384-well assay protocol.

The QuantiGene Plex Automation Setup Kit enables setup, testing, and validation of instruments to automate crucial steps of the QuantiGene Plex 384-well assay protocol, such as liquid handling and plate washing, before running an actual assay with valuable sample. The user guide describes how to adjust and optimize an automated magnetic plate washer as needed during the QuantiGene Plex 384-well assay workflow in order to ensure optimal performance and avoid bead loss.

Each kit provides six optimization tests for automated magnetic plate washing.

TRUSTED_SUSTAINABILITY

Specifications

Channel Classification Virology
Format 384-well
For Use With (Equipment) FLEXMAP 3D™ INTELLIFLEX™
Content And Storage Virology
Product Line QuantiGene™ Plex
Product Type Automation Setup Kit
Quantity 6 Tests
Shipping Condition Wet Ice
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.