Learn More
Titin Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27867-1-AP-20UL
This item is not returnable.
View return policy
Description
The Titin antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human Titin. This antibody recognizes human, mouse antigen. The Titin antibody has been validated for the following applications: IF, IHC, ELISA analysis.
Specifications
| Titin | |
| Polyclonal | |
| Unconjugated | |
| 20 μL | |
| 7273 | |
| IgG |
| Immunohistochemistry, Immunofluorescence | |
| 333 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Mouse |