missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Titin Polyclonal Antibody

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA552379

Product Code. 15740331

  • £438.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: GTVSTSCYLA VQVSEEFEKE TTAVTEKFTT EEKRFVESRD VVMTDTSLTE EQAGPGEPAA PYFITKPVVQ KLVEGGSVVF GCQVGGNPKP HVYWKKSGVP LTTGYRYKVS YNKQTGECKL VISMTFADDA GEYT Highest antigen sequence identity to the following orthologs: Mouse - 93%, Rat - 27%.

Monoclonal Anti-Titin can be used for study of the elastic filaments within sarcomeric structures. It is also useful as a differentiation marker in the separation of rhabdomyosarcomas from other muscle tumors. By indirect immunofluorescence the antibody displays a typical striated staining pattern on frozen sections of chicken skeletal and cardiac muscle tissues. Stains the region of the A-I junction by indirect immunofluorescence. It shows a decoration line 0.05 mm from the end of the A band in electron micro-graphs. In immunoblotting using total extracts of chicken breast muscle the antibody reacts specifically with both bands of the titin doublet, and showed no reaction with nebulin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Titin
Polyclonal
Unconjugated
TTN
1100001C23Rik; 2.7.11.1; 2310036G12Rik; 2310057K23Rik; 2310074I15Rik; AF006999; AV006427; CMD1G; CMH9; CMPD4; Connectin; connectin/titin; D330041I19Rik; D830007G01Rik; DKFZp451N061; EOMFC; FLJ26020; FLJ26409; FLJ32040; FLJ34413; FLJ39564; FLJ43066; HMERF; L56; LGMD2J; mdm; MYLK5; rhabdomyosarcoma antigen MU-RMS-40.14; shru; structural muscle protein titin; titin; titin protein homolog; titin sequence3; TMD; TTN
Rabbit
Antigen Affinity Chromatography
RUO
7273
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
0.10 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q8WZ42
TTN
Recombinant protein corresponding to Human Titin. Recombinant protein control fragment (Product #RP-89761).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.