missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Titin Antibody (T11), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 3 publications
Brand: Novus Biologicals NB600-1206
This item is not returnable.
View return policy
Description
Titin Monoclonal specifically detects Titin in Human, Mouse, Rat, Avian, Chicken, Fish, Insect, Mammal, Reptile samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Electron Microscopy.
Specifications
| Titin | |
| Monoclonal | |
| Unconjugated | |
| Ascites with 0.1% Sodium Azide | |
| TTN | |
| Full length native protein (purified) (Chicken breast muscle). | |
| 0.1 mL | |
| Cancer, Cytoskeleton Markers | |
| 7273 | |
| Human, Mouse, Rat, Avian, Chicken, Fish, Insect, Mammalia, Reptile | |
| Ascites |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| T11 | |
| Western Blot 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:1000, Immunohistochemistry-Frozen 1:10-1:500, Electron Microscopy 1:10-1:500 | |
| cardiomyopathy, dilated 1G (autosomal dominant), CMD1G, CMH9, CMPD4, Connectin, DKFZp451N061, EC 2.7.11.1, EOMFC, FLJ32040, FLJ34413, FLJ39564, FLJ43066, HMERF, LGMD2JFLJ26020, MPRM, Rhabdomyosarcoma antigen MU-RMS-40.14, titin, TMDFLJ26409 | |
| Mouse | |
| Unpurified | |
| RUO | |
| Primary | |
| The antibody localizes titin (connectin) in skeletal and heart muscle of a wide variety of species from cold-blooded vertebrates to human. The antibody does not cross-react with nebulin. Likewise, it does not react with smooth muscle, non-muscle tissues, or cultured cells. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction