missing translation for 'onlineSavingsMsg'
Learn More

Titanium wire, 0.25mm (0.01in) dia, 99.99% (metals basis), Thermo Scientific Chemicals

Catalog Number 11365858
Click to view available options
Quantity:
1 m
25 m
5 m
This item is not returnable. View return policy
7440-32-6
Ti
47.87
MFCD00011264,MFCD00151301
RTAQQCXQSZGOHL-UHFFFAOYSA-N
oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
23963
CHEBI:33341
titanium
[Ti]
This item is not returnable. View return policy
7440-32-6
Ti
47.87
MFCD00011264,MFCD00151301
RTAQQCXQSZGOHL-UHFFFAOYSA-N
oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
23963
CHEBI:33341
titanium
[Ti]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 1 m
Percent Purity 99.99%
Odor Odorless
Weight ≈0.22g/m
Form Wire
Assay (metals basis)
Chemical Name or Material Titanium wire, 0.25mm (0.01 in.) dia., Temper: as drawn

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'