missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Titanium, Ti 6-7, R56700, 65g, ARMI∣MBH
Click to view available options
Quantity:
65 g
Unit Size:
65g
Description
Titanium, Ti 6-7, R56700, 65g
Specifications
Specifications
| Chemical Name or Material | Molybdenum/Manganese/Hydrogen/Zirconium/Nitrogen/Copper/Tantalum/Vanadium/Carbon/Tin/Nickel/Chromium/Silicon/Oxygen/Iron/Aluminium/Niobium/Titanium/Tungsten/Magnesium/Yttrium/Boron/Cobalt/Phosphorus/Palladium |
| CAS | 7440-32-6 |
| Quantity | 65 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Molybdenum 0.0009 %/Manganese 0.0012 %/Hydrogen 0.0018 %/Zirconium 0.003 %/Nitrogen 0.0033 %/Copper 0.0039 %/Tantalum 0.004 %/Vanadium 0.005 %/Carbon 0.007 %/Tin 0.007 %/Nickel 0.0081 %/Chromium 0.0097 %/Silicon 0.018 %/Oxygen 0.162 %/Iron 0.191 %/Aluminium 5.96 %/Niobium 6.86 %/Titanium 86.71 %/Tungsten (<0.002) %/Magnesium (0.0004) %/Yttrium (0.0004) %/Boron (0.0008) %/Cobalt (0.001) %/Phosphorus (0.001) %/Palladium (0.002) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction