missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Titanium, Ti 6-4, Grade 5, R56400, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Titanium, Ti 6-4, Grade 5, R56400, Ø38x3mm
Specifications
Specifications
| Chemical Name or Material | Tin/Hydrogen/Molybdenum/Nickel/Silicon/Chromium/Nitrogen/Carbon/Oxygen/Iron/Vanadium/Aluminium/Cadmium/Palladium/Arsenic/Bismuth/Calcium/Phosphorus/Selenium/Yttrium/Boron/Hafnium/Lead/Magnesium/Ruthenium/Zinc/Zirconium/Antimony/Tantalum/Tungsten/Cobalt/Manganese/Copper/Niobium/Titanium |
| CAS | 7440-32-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Tin 0.003 %/Hydrogen 0.0031 %/Molybdenum 0.004 %/Nickel 0.011 %/Silicon 0.011 %/Chromium 0.013 %/Nitrogen 0.025 %/Carbon 0.034 %/Oxygen 0.181 %/Iron 0.19 %/Vanadium 4.08 %/Aluminium 6.52 %/Cadmium (<0.0010) %/Palladium (<0.0010) %/Arsenic (<0.0050) %/Bismuth (<0.0050) %/Calcium (<0.0050) %/Phosphorus (<0.0050) %/Selenium (<0.0050) %/Yttrium (<0.0050) %/Boron (<0.0100) %/Hafnium (<0.0100) %/Lead (<0.0100) %/Magnesium (<0.0100) %/Ruthenium (<0.0100) %/Zinc (<0.0100) %/Zirconium (<0.0100) %/Antimony (<0.0200) %/Tantalum (0.0002) %/Tungsten (0.0002) %/Cobalt (0.0004) %/Manganese (0.001) %/Copper (0.002) %/Niobium (0.004) %/Titanium (89) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction