missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Titanium, Ti 5-2.5, Grade 6, R54520, 65g, ARMI∣MBH
Click to view available options
Quantity:
65 g
Unit Size:
65g
Description
Titanium, Ti 5-2.5, Grade 6, R54520, 65g
Specifications
Specifications
| Chemical Name or Material | Boron/Manganese/Niobium/Molybdenum/Copper/Nickel/Chromium/Silicon/Carbon/Nitrogen/Hydrogen/Vanadium/Zirconium/Oxygen/Iron/Tin/Aluminium/Yttrium/Cobalt/Magnesium/Palladium/Phosphorus/Tungsten/Ruthenium/Tantalum/Titanium |
| CAS | 7440-32-6 |
| Quantity | 65 g |
| Form | Pieces |
| Concentration or Composition (by Analyte or Components) | Boron 0.0012 %/Manganese 0.0036 %/Niobium 0.005 %/Molybdenum 0.0063 %/Copper 0.0068 %/Nickel 0.0089 %/Chromium 0.01 %/Silicon 0.01 %/Carbon 0.0128 %/Nitrogen 0.013 %/Hydrogen 0.0139 %/Vanadium 0.057 %/Zirconium 0.059 %/Oxygen 0.155 %/Iron 0.196 %/Tin 2.52 %/Aluminium 5.21 %/Yttrium (0.00018) %/Cobalt (0.001) %/Magnesium (0.001) %/Palladium (0.0011) %/Phosphorus (0.0011) %/Tungsten (0.0021) %/Ruthenium (0.003) %/Tantalum (0.0104) %/Titanium (91.9) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction