missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Titanium, Ti 3-2.5, Grade 9, R56320, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Titanium, Ti 3-2.5, Grade 9, R56320, Ø32x19mm
Specifications
Specifications
| Chemical Name or Material | Boron/Copper/Molybdenum/Tin/Nitrogen/Silicon/Carbon/Chromium/Nickel/Oxygen/Iron/Vanadium/Aluminium/Cobalt/Hydrogen/Manganese/Ruthenium/Tungsten/Yttrium/Palladium/Zirconium/Niobium/Titanium/Phosphorus |
| CAS | 7440-32-6 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Boron 0.0003 %/Copper 0.0025 %/Molybdenum 0.003 %/Tin 0.005 %/Nitrogen 0.006 %/Silicon 0.007 %/Carbon 0.012 %/Chromium 0.016 %/Nickel 0.018 %/Oxygen 0.084 %/Iron 0.18 %/Vanadium 2.51 %/Aluminium 3.05 %/Cobalt (0.0004) %/Hydrogen (0.0005) %/Manganese (0.001) %/Ruthenium (0.001) %/Tungsten (0.001) %/Yttrium (0.001) %/Palladium (0.002) %/Zirconium (0.003) %/Niobium (0.005) %/Titanium (94.1) %/Phosphorus <0.003 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction