missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Titanium, Ti 3-2.5, Grade 9, R56320, ARMI∣MBH
Click to view available options
Unit Size:
Each
Description
Titanium, Ti 3-2.5, Grade 9, R56320, Ø32x19mm
Specifications
Specifications
| Chemical Name or Material | Boron/Copper/Molybdenum/Zirconium/Tin/Nitrogen/Silicon/Carbon/Chromium/Nickel/Oxygen/Iron/Vanadium/Aluminium/Hydrogen/Ruthenium/Tantalum/Yttrium/Tungsten/Manganese/Palladium/Niobium/Titanium/Cobalt/Phosphorus |
| CAS | 7440-32-6 |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Boron 0.0003 %/Copper 0.0028 %/Molybdenum 0.003 %/Zirconium 0.003 %/Tin 0.005 %/Nitrogen 0.0051 %/Silicon 0.008 %/Carbon 0.011 %/Chromium 0.016 %/Nickel 0.018 %/Oxygen 0.083 %/Iron 0.185 %/Vanadium 2.5 %/Aluminium 3.02 %/Hydrogen (0.0005) %/Ruthenium (0.0005) %/Tantalum (0.0005) %/Yttrium (0.0005) %/Tungsten (0.001) %/Manganese (0.002) %/Palladium (0.002) %/Niobium (0.003) %/Titanium (94.2) %/Cobalt <0.001 %/Phosphorus <0.004 % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction