missing translation for 'onlineSavingsMsg'
Learn More

Titanium sputtering target, 50.8mm (2.0in) dia x 6.35mm (0.250in) thick, 99.7% (metals basis)

Product Code. 15404835
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 15404835

Brand: Thermo Scientific Alfa Aesar 040903.KS

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7440-32-6 | Ti | 47.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 Ea.
CAS 7440-32-6
Odor Odorless
Molecular Formula Ti
MDL Number MFCD00011264,MFCD00151301
Synonym oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
InChI Key RTAQQCXQSZGOHL-UHFFFAOYSA-N
SMILES [Ti]
IUPAC Name titanium
Molecular Weight (g/mol) 47.87
PubChem CID 23963
ChEBI CHEBI:33341
Percent Purity 99.7%
Assay (metals basis)
Chemical Name or Material Titanium sputtering target, 50.8mm (2.0 in.) dia. x 6.35mm (0.250 in.) thick
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.