missing translation for 'onlineSavingsMsg'
Learn More

Titanium slug, 9.5mm (0.37 in.) dia. x 9.5mm (0.37 in.) length, 99.9+% (metals basis), Thermo Scientific Chemicals

Product Code. 11365838
Change view
Click to view available options
Quantity:
1 Slug
25 Slugs
5 Slugs
Unit Size:
Each
Pack of 25
Pack of 5
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11365838 25 Slugs Pack of 25
11345838 1 Slug Each
11355838 5 Slugs Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11365838 Supplier Thermo Scientific Alfa Aesar Supplier No. 010381.QK
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-32-6
Molecular Formula Ti
Molecular Weight (g/mol) 47.87
MDL Number MFCD00011264,MFCD00151301
InChI Key RTAQQCXQSZGOHL-UHFFFAOYSA-N
Synonym oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
PubChem CID 23963
ChEBI CHEBI:33341
IUPAC Name titanium
SMILES [Ti]

Specifications

Quantity 25 Slugs
Odor Odorless
Weight ≈3g/slug
Form Slug
Percent Purity ≥99.9%
Assay (metals basis)
Chemical Name or Material Titanium slug, 9.5mm (0.37 in.) dia. x 9.5mm (0.37 in.) length

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.