missing translation for 'onlineSavingsMsg'
Learn More

Titanium silicide, 99.5% (metals basis)

Product Code. 11372178
Change view
Click to view available options
Quantity:
50 g
250 g
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11372178 250 g 250g
11362178 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11372178 Supplier Thermo Scientific Alfa Aesar Supplier No. 013087.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12067-57-1 | Si3Ti5 | 323.59 g/mol

It is used as source, gate and drain contacts and local interconnections in CMOS integrated circuits. it is used in the C54 phase.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12067-57-1
Molecular Formula Si3Ti5
Molecular Weight (g/mol) 323.59
MDL Number MFCD01310208
InChI Key GNKTZDSRQHMHLZ-UHFFFAOYSA-N
Synonym silicon; titanium, titanium silicide ti5si3, pentatitanium trisilicide, titanium silicide,-325 mesh
PubChem CID 6336553
IUPAC Name silicon;titanium
SMILES [Si].[Si].[Si].[Ti].[Ti].[Ti].[Ti].[Ti]

Specifications

Melting Point 2130°C
Physical Form Powder
Quantity 250 g
Assay Percent Range 99.5%, metal basis
Solubility Information Soluble in hydrofluoric acid. Insoluble in water.
Formula Weight 323.76
Chemical Name or Material Titanium Silicide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.