missing translation for 'onlineSavingsMsg'
Learn More

Titanium powder, dehydrided, -325 mesh, 99.99% (metals basis), Thermo Scientific Chemicals

£167.00 - £483.00

Chemical Identifiers

CAS 7440-32-6
Molecular Formula Ti
Molecular Weight (g/mol) 47.87
MDL Number MFCD00011264,MFCD00151301
InChI Key RTAQQCXQSZGOHL-UHFFFAOYSA-N
Synonym oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
PubChem CID 23963
ChEBI CHEBI:33341
IUPAC Name titanium
SMILES [Ti]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11364197
View Documents Promotion Details
Thermo Scientific Alfa Aesar
036679.14
25 g
£167.00
25g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11354197
View Documents Promotion Details
Thermo Scientific Alfa Aesar
036679.22
100 g
£483.00
100g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7440-32-6
47.87
RTAQQCXQSZGOHL-UHFFFAOYSA-N
23963
titanium
Ti
MFCD00011264,MFCD00151301
oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
CHEBI:33341
[Ti]

Specifications

Gray
Dark Gray powder
Ti
oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
[Ti]
47.87
CHEBI:33341
Odorless
Titanium
7440-32-6
99.99% (Metals basis)
MFCD00011264,MFCD00151301
RTAQQCXQSZGOHL-UHFFFAOYSA-N
titanium
23963
-325 Mesh
Dehydrided Powder
Videos
Safety and Handling

Safety and Handling

P210-P222-P280-P335+P334-P370+P378q

H250-H500

missing translation for 'dotInformation' : DOT Class: 4.2, Packaging Group: II

missing translation for 'einecsNumber' : 231-142-3

missing translation for 'rtecsNumber' : XR1700000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Titanium powder

Signal Word
  • Danger

Hazard Category
  • Pyrophoric solid Category 1
  • Self-heating substance Category 1

Hazard Statement
  • H250-Catches fire spontaneously if exposed to air.
  • H251-Self-heating: may catch fire.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P222-Do not allow contact with air.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P235+P410-Keep cool. Protect from sunlight.
  • P422-Store contents under inert gas.
  • P420-Store away from other materials.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only