missing translation for 'onlineSavingsMsg'
Learn More

Titanium powder, dehydrided, -325 mesh, 99.99% (metals basis)

Product Code. 11354197
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11354197 100 g 100g
11364197 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11354197 Supplier Thermo Scientific Alfa Aesar Supplier No. 036679.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7440-32-6 | Ti | 47.87 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-32-6
Molecular Formula Ti
Molecular Weight (g/mol) 47.87
MDL Number MFCD00011264,MFCD00151301
InChI Key RTAQQCXQSZGOHL-UHFFFAOYSA-N
Synonym oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
PubChem CID 23963
ChEBI CHEBI:33341
IUPAC Name titanium
SMILES [Ti]

Specifications

Color Gray
Appearance Dark Gray powder
Quantity 100 g
Mesh Size -325 Mesh
Odor Odorless
Assay Percent Range 99.99% (Metals basis)
Form Dehydrided Powder
Chemical Name or Material Titanium
compliance-icons
Product Identifier
  • Titanium powder
Signal Word
  • Danger
Hazard Category
  • Pyrophoric solid Category 1
  • Self-heating substance Category 1
Hazard Statement
  • H250-Catches fire spontaneously if exposed to air.
  • H251-Self-heating: may catch fire.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P222-Do not allow contact with air.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P235+P410-Keep cool. Protect from sunlight.
  • P422-Store contents under inert gas.
  • P420-Store away from other materials.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.