missing translation for 'onlineSavingsMsg'
Learn More

Titanium powder, -325 mesh, 99.5% (metals basis), Thermo Scientific Chemicals

Catalog Number 11359248
Click to view available options
Quantity:
100 g
500 g
2 x 1 kg
7440-32-6
Ti
47.87
MFCD00011264,MFCD00151301
RTAQQCXQSZGOHL-UHFFFAOYSA-N
oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
23963
CHEBI:33341
titanium
[Ti]
7440-32-6
Ti
47.87
MFCD00011264,MFCD00151301
RTAQQCXQSZGOHL-UHFFFAOYSA-N
oremet, alloy, cp, vt1, c.p., contimet 30, jistp28, titanium-125, ii hydride, titan vt 1-1
23963
CHEBI:33341
titanium
[Ti]

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Silver to Gray
Appearance Silver Gray powder
Density 4.506g/cm3
Assay Percent Range 99.5% (Metals basis)
Quantity 100 g
Mesh Size -325 Mesh
Odor Odorless
Form Powder
Chemical Name or Material Titanium

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'