missing translation for 'onlineSavingsMsg'
Learn More

Titanium oxide, 99.9% (metals basis)

Product Code. 11349057
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11349057 25 g 25g
11339057 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11349057 Supplier Thermo Scientific Alfa Aesar Supplier No. 039000.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Useful for photo voltaic applications. It plays an important role as photocatalyst under visible light radiation. Treated titanium oxide nanotubes play a significant part in the development of nano-scale hydroxy apatite. It is too utilized as catalysts. | CAS: 12065-65-5 | Ti3O5

Nanocrystalline titanium oxide electrodes is useful for photo voltaic applications. It plays an important role as photocatalyst under visible light radiation. Treated titanium oxide nanotubes play a significant part in the development of nano-scale hydroxy apatite. It is too utilized as catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12065-65-5
Molecular Formula Ti3O5
MDL Number MFCD00210650
Synonym Trititanium pentoxide

Specifications

Melting Point 1800°C
Physical Form Pieces
Quantity 25 g
Solubility Information Soluble in hot concentrated sulfuric acid,hydrofluoric acid and alkali.
Percent Purity 99.9%
Packaging Plastic bottle
Specific Gravity 4.2
Density 4.2
Chemical Name or Material Titanium oxide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.