missing translation for 'onlineSavingsMsg'
Learn More

Titanium nitride, 99.7% (metals basis)

Product Code. 11372458
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
This item is not returnable. View return policy

Product Code. 11372458

Brand: Thermo Scientific Alfa Aesar 014510.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 25583-20-4 | NTi | 61.87 g/mol

Titanium nitride is a hard ceramic material used as a coating on titanium alloys, steel, carbide and aluminum components. It finds application in jewelry and high-performance ceramic materials due to its gold colored surface and in medical devices such as scalpel blades and orthopedic bone saw blades. Further, it is used as a conductive barrier in microelectronics and electrodes in bioelectronic applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25583-20-4
Molecular Formula NTi
Molecular Weight (g/mol) 61.87
MDL Number MFCD00049596
InChI Key NRTOMJZYCJJWKI-UHFFFAOYSA-N
Synonym titanium nitride, titanium nitride tin, nitridotitanium, unii-6rw464feff, titannitrid, titaniumylidyneamine, titanium mononitride, titanium nitride, <3 mum, 6rw464feff, titanium nitride tin grade a
PubChem CID 93091
IUPAC Name azanylidynetitanium
SMILES N#[Ti]

Specifications

Melting Point 2950°C
Physical Form Powder
Quantity 250 g
Assay Percent Range 99.7% (metal basis)
Solubility Information Insoluble in water.
Formula Weight 61.91
Density 5.22
Chemical Name or Material Titanium nitride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.