missing translation for 'onlineSavingsMsg'
Learn More

Titanium nitride, 99.7% (metals basis)

Product Code. 11382458
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11382458 50 g 50g
11372458 250 g 250g
11392458 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11382458 Supplier Thermo Scientific Alfa Aesar Supplier No. 014510.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 25583-20-4 | NTi | 61.87 g/mol

Titanium nitride is a hard ceramic material used as a coating on titanium alloys, steel, carbide and aluminum components. It finds application in jewelry and high-performance ceramic materials due to its gold colored surface and in medical devices such as scalpel blades and orthopedic bone saw blades. Further, it is used as a conductive barrier in microelectronics and electrodes in bioelectronic applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 25583-20-4
Molecular Formula NTi
Molecular Weight (g/mol) 61.87
MDL Number MFCD00049596
InChI Key NRTOMJZYCJJWKI-UHFFFAOYSA-N
Synonym titanium nitride, titanium nitride tin, nitridotitanium, unii-6rw464feff, titannitrid, titaniumylidyneamine, titanium mononitride, titanium nitride, <3 mum, 6rw464feff, titanium nitride tin grade a
PubChem CID 93091
IUPAC Name azanylidynetitanium
SMILES N#[Ti]

Specifications

Melting Point 2950°C
Physical Form Powder
Quantity 50 g
Assay Percent Range 99.7% (metal basis)
Solubility Information Insoluble in water.
Formula Weight 61.91
Density 5.22
Chemical Name or Material Titanium nitride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.