missing translation for 'onlineSavingsMsg'
Learn More

Titanium Nickel powder composite, -325 mesh

Product Code. 11339259
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
Unit Size:
100g
10g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11339259 500 g 500g
11329259 10 g 10g
11349259 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11339259 Supplier Thermo Scientific Alfa Aesar Supplier No. 088384.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

A 70% titanium, 30% nickel composite powder strained through -325 mesh. Also used in alloy powder, bone staples and antioxidant coating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Titanium Nickel powder composite
Avg. Mol. Wt. or Mol. Wt. Range Ti:Ni; 70:30 wt%
Quantity 500 g
Mesh Size −325 Mesh
Sensitivity Air Sensitive
Form Powder
MDL Number MFCD02091734
UN Number UN3089
Solubility Information Insoluble in water.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.