missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) tert-butoxide, 98%, Thermo Scientific Chemicals

Catalog Number p-200653
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
3087-39-6
C16H36O4Ti
340.33
MFCD00040554
GRWPYGBKJYICOO-UHFFFAOYSA-N
unii-px6yr1s99s, px6yr1s99s, titanium iv tert-butoxide, titanium tetra-tert-butoxide, titanium tert-butoxide, titanum tetra t-butoxide, tetra-tert-butoxytitanium iv, titanium 4+ 2-methylpropan-2-olate
6451515
[Ti+4].CC(C)(C)[O-].CC(C)(C)[O-].CC(C)(C)[O-].CC(C)(C)[O-]
This item is not returnable. View return policy
3087-39-6
C16H36O4Ti
340.33
MFCD00040554
GRWPYGBKJYICOO-UHFFFAOYSA-N
unii-px6yr1s99s, px6yr1s99s, titanium iv tert-butoxide, titanium tetra-tert-butoxide, titanium tert-butoxide, titanum tetra t-butoxide, tetra-tert-butoxytitanium iv, titanium 4+ 2-methylpropan-2-olate
6451515
[Ti+4].CC(C)(C)[O-].CC(C)(C)[O-].CC(C)(C)[O-].CC(C)(C)[O-]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Lewis acid
TRUSTED_SUSTAINABILITY
Color Colorless
Density 0.8893g/mL
Boiling Point 160.0°C to 165.0°C (15.0 mmHg)
Flash Point 47°C
Assay Percent Range 97.5% min
Packaging Glass bottle
Linear Formula Ti[OC(CH3)3]4
Quantity 5 g
Beilstein 01, IV, 1613
Specific Gravity 0.8893
IUPAC Name titanium(4+) tetrakis(2-methylpropan-2-olate)
Formula Weight 340.33
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Titanium(IV) tert-butoxide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Titanium(IV)tert-butoxide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'