missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) sulfide, 99.8% (metals basis)

Product Code. 11347557
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11347557

Brand: Thermo Scientific Alfa Aesar 012826.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 12039-13-3 | S2Ti | 111.99 g/mol

Titanium(IV) sulfide is used in solid lubricant, cathode material in rechargeable batteries, corrosion resistance alloys, solid-state lithium batteries, hybrid electric vehicles and plug-in electric vehicles. It is used in the synthesis of nanoclusters, nanodisks and nanotubes. It has high electrical conductivity. Titanium electrodes are used in chloralkali electrolysis, electrodialysis, galvanic technology, light bulbs, and bone pins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12039-13-3
Molecular Formula S2Ti
Molecular Weight (g/mol) 111.99
MDL Number MFCD00049598
InChI Key CFJRPNFOLVDFMJ-UHFFFAOYSA-N
Synonym titanium sulfide tis2, titanium disulfide, titanium iv sulfide, bis sulfanylidene titanium, titanium disulphide, titansulfid, titanium sulfide, tis2, titanium iv sulfide-ti, titanium disulfide, crystal
PubChem CID 61544
SMILES S=[Ti]=S

Specifications

Physical Form Powder
Quantity 25 g
Assay Percent Range 99.8%, metal basis
Solubility Information Titanium(IV) sulfide is insoluble in water and moderately acid soluble.
IUPAC Name bis(sulfanylidene)titanium
Formula Weight 112.03
Sensitivity Moisture sensitive
Chemical Name or Material Titanium(IV) Sulfide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.