missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) sulfide, 99.8% (metals basis), Thermo Scientific Chemicals

Product Code. 11357557
Click to view available options
Quantity:
5 g
25 g

Titanium(IV) sulfide is used in solid lubricant, cathode material in rechargeable batteries, corrosion resistance alloys, solid-state lithium batteries, hybrid electric vehicles and plug-in electric vehicles. It is used in the synthesis of nanoclusters, nanodisks and nanotubes. It has high electrical conductivity. Titanium electrodes are used in chloralkali electrolysis, electrodialysis, galvanic technology, light bulbs, and bone pins.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12039-13-3
Molecular Formula S2Ti
Molecular Weight (g/mol) 111.99
MDL Number MFCD00049598
InChI Key CFJRPNFOLVDFMJ-UHFFFAOYSA-N
Synonym titanium sulfide tis2, titanium disulfide, titanium iv sulfide, bis sulfanylidene titanium, titanium disulphide, titansulfid, titanium sulfide, tis2, titanium iv sulfide-ti, titanium disulfide, crystal
PubChem CID 61544
SMILES S=[Ti]=S

Specifications

Physical Form Powder
Quantity 5 g
Assay Percent Range 99.8%, metal basis
Solubility Information Titanium(IV) sulfide is insoluble in water and moderately acid soluble.
IUPAC Name disulfanylidenetitanium
Formula Weight 112.03
Sensitivity Moisture sensitive
Chemical Name or Material Titanium(IV) Sulfide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'