missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) Sulfate, Technical, Solution ≈15% (w/v)

Product Code. 10326250 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Plastic Coated Glass bottle
Unit Size:
1 L
Product Code. Quantity Packaging unitSize
10326250 1 L Plastic Coated Glass bottle 1 L
1 options
This item is not returnable. View return policy

Product Code. 10326250

Brand: Fisher Chemical T/1800/PB15

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 13693-11-3
Color Colorless
Physical Form Liquid
Quantity 1 L
MDL Number 168084
Packaging Plastic Coated Glass bottle
compliance-icons
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Sulfuric acid, Titanium sulfate (Ti(SiO4)2)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.