missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) oxide sulfate sulfuric acid hydrate

Product Code. 11344507
Change view
Click to view available options
Quantity:
500 g
2 kg
Unit Size:
2kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11344507 2 kg 2kg
11354507 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11344507 Supplier Thermo Scientific Alfa Aesar Supplier No. 039591.A3
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 123334-00-9 | TiOSO4.xH2O + H2SO4

Titanium(IV) oxide sulfate sulfuric acid hydrate is widely used as a pigment in paints and dyes. It is also useful in the preparation of stable amorphous titanium oxide hollow microspheres.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123334-00-9
Molecular Formula TiOSO4.xH2O + H2SO4
MDL Number MFCD00011270
Synonym Titanyl sulfate, Titanium(IV) oxysulfate-sulfuric acid

Specifications

Color White
Physical Form Powder
Quantity 2 kg
Solubility Information Soluble in water.
Chemical Name or Material Titanium(IV) oxide sulfate sulfuric acid hydrate
compliance-icons
Product Identifier
  • Titanium(IV) oxide sulfate sulfuric acid hydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 A
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Titanium(IV) oxide sulfate sulfuric acidhydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.