missing translation for 'onlineSavingsMsg'
Learn More

Titanium(IV) oxide sulfate sulfuric acid hydrate, Thermo Scientific Chemicals

Catalog Number 11354507
Click to view available options
Quantity:
500 g
2 kg
This item is not returnable. View return policy
123334-00-9
TiOSO4.xH2O + H2SO4
MFCD00011270
Titanyl sulfate, Titanium(IV) oxysulfate-sulfuric acid
This item is not returnable. View return policy
123334-00-9
TiOSO4.xH2O + H2SO4
MFCD00011270
Titanyl sulfate, Titanium(IV) oxysulfate-sulfuric acid

Titanium(IV) oxide sulfate sulfuric acid hydrate is widely used as a pigment in paints and dyes. It is also useful in the preparation of stable amorphous titanium oxide hollow microspheres.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color White
Physical Form Powder
Quantity 500 g
Solubility Information Soluble in water.
Chemical Name or Material Titanium(IV) oxide sulfate sulfuric acid hydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'